Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionicotinamide Adenine Dinucleotide oxidized form 95.0+%, TCI America™
SDP

Catalog No. T2979100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2979100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2979100MG Supplier TCI America Supplier No. T2979100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4090-29-3
Molecular Formula C21H27N7O13P2S
Molecular Weight (g/mol) 679.491
MDL Number MFCD00079609
InChI Key UQYPZLRUJKCREN-WWRWIPRPSA-N
Synonym Thionicotinamide-DPN
PubChem CID 91972082
IUPAC Name [[(2R,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,4S,5R)-5-(3-carbamothioylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=S)N

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 679.49
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Thionicotinamide Adenine Dinucleotide oxidized form [for Biochemical Research]
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.