Learn More
Medchemexpress LLC Thionin acetate | 78338-22-4 | 93.00% | 500 MG
SDP
Catalog No. 5000409705
Encompass_Preferred
Thionin acetate (Thionine acetate) is a compound found in the seeds, stems, roots, and leaves of many plant species with antibacterial activity. It is also a metachromatic cationic histological dye widely used in biological staining.
  • Can be used for tissue staining, such as staining brainstem slices, to observe and analyze brainstem structure and determine the location of the preBötC and characteristics of related regions.
  • The staining process involves treating tissue sections with ethanol and distilled water, then immersing them in Thionin acetate solution, followed by washing and clearing in xylene for observation and analysis.
Catalog No. 50-004-09705 Supplier Medchemexpress LLC Supplier No. HYD0955500MG
Thionin acetate (Thionine acetate) is a compound found in the seeds, stems, roots, and leaves of many plant species with antibacterial activity. It is also a metachromatic cationic histological dye widely used in biological staining.
  • Can be used for tissue staining, such as staining brainstem slices, to observe and analyze brainstem structure and determine the location of the preBötC and characteristics of related regions.
  • The staining process involves treating tissue sections with ethanol and distilled water, then immersing them in Thionin acetate solution, followed by washing and clearing in xylene for observation and analysis.