Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionin, Certified, Thermo Scientific™

Catalog No. AC420880250
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420880250 25 g
AC420880100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420880250 Supplier Thermo Scientific Chemicals Supplier No. 420880250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-64-6
Molecular Formula C12H10ClN3S
Molecular Weight (g/mol) 263.743
InChI Key PVPBBTJXIKFICP-UHFFFAOYSA-N
PubChem CID 65043
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;chloride
SMILES C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2.[Cl-]

Specifications

Absorption Ratio 1.15 to 1.57
Dye Content 85% min.
Merck Index 15, 9499
Infrared Spectrum Authentic
Lambda Max 598 to 600nm (in water)
Formula Weight 263.75
Color Black to Green
Quantity 25 g
Physical Form Powder or Crystals
Chemical Name or Material Thionin
EINECSNumber 209-470-3
RTECSNumber SN5425000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.