Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Thionin Solution
SCI_ED

Catalog No. S25817 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25817 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25817
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 581-64-6 , 7732-18-5
Molecular Formula C12H10ClN3S
Molecular Weight (g/mol) 263.743
InChI Key PVPBBTJXIKFICP-UHFFFAOYSA-N
Synonym C.I. 52000, Thionine
PubChem CID 65043
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;chloride
SMILES C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2.[Cl-]

Specifications

Packaging Solution
Assay Percent Range 0.006
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Thionin Solution
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.