Learn More
Medchemexpress LLC Thionine acetate | 78338-22-4 | 93.0% | 1 G
SDP
Catalog No. 5000409744
Encompass_Preferred
Thionin acetate is a compound found in various plant species, known for its antibacterial activity. It also serves as a metachromatic cationic histological dye, widely used in biological staining for observing and analyzing tissue structures like brainstem slices.
  • Possesses antibacterial activity
  • Functions as a metachromatic cationic histological dye
  • Used for biological staining
  • Suitable for tissue staining, including brainstem slices
Catalog No. 50-004-09744 Supplier Medchemexpress LLC Supplier No. HYD09551G
Thionin acetate is a compound found in various plant species, known for its antibacterial activity. It also serves as a metachromatic cationic histological dye, widely used in biological staining for observing and analyzing tissue structures like brainstem slices.
  • Possesses antibacterial activity
  • Functions as a metachromatic cationic histological dye
  • Used for biological staining
  • Suitable for tissue staining, including brainstem slices