Learn More
Sigma Aldrich Fine Chemicals Biosciences Thionyl chloride, 7719-09-7, MFCD00011449, 100mL
Catalog No. 501657225
Encompass
Linear Formula SOCl2, ≥99%, Molecular Weight 118.97, bp 79 °C (lit.), mp −105 °C (lit.), ReagentPlus
Catalog No. 50-165-7225 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 230464100ML
Linear Formula SOCl2, ≥99%, Molecular Weight 118.97, bp 79 °C (lit.), mp −105 °C (lit.), ReagentPlus