Learn More
Sigma Aldrich Fine Chemicals Biosciences Thionyl chloride ReagentPl
Catalog No. 502897763
Encompass
Thionyl chloride is an inorganic acid chloride mainly used to prepare carboxylic acid chlorides from carboxylic acids.
Catalog No. 50-289-7763 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2304642.5L
Thionyl chloride is an inorganic acid chloride mainly used to prepare carboxylic acid chlorides from carboxylic acids.