Learn More
TARGETMOL CHEMICALS INC Thioperamide maleate 10MG
SDP
Catalog No. 502880458
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg and bulk. Please contact Fisher for quotes. Thioperamide maleate (MR-12842 maleate) is an effective and selective H3 receptor antagonist (Ki = 4.3 nM) that inhibits [3H]histamine synthesis (Ki = 31 nM). Purity 99.88%
Catalog No. 50-288-0458 Supplier TARGETMOL CHEMICALS INC Supplier No. T4124710MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg and bulk. Please contact Fisher for quotes. Thioperamide maleate (MR-12842 maleate) is an effective and selective H3 receptor antagonist (Ki = 4.3 nM) that inhibits [3H]histamine synthesis (Ki = 31 nM). Purity 99.88%