Learn More
Medchemexpress LLC Thioperamide maleate | 148440-81-7 | >98.0% | 10 MG
SDP
Catalog No. 5000231577
Encompass_Preferred
Thioperamide maleate is the maleate salt of thioperamide, a selective histamine H3 receptor antagonist used as a pharmacological tool to study histaminergic signaling. It is brain-penetrant and orally active, with a reported Ki of approximately 4.3 nM for H3 receptor inhibition.
  • Selective H3 receptor antagonist with Ki ≈ 4.3 nM.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • Provided as a solid, commonly supplied in 10 mg vials for research use.
  • High purity (>98%) as determined by vendor quality control.
Catalog No. 50-002-31577 Supplier Medchemexpress LLC Supplier No. HY12206A10MG
Thioperamide maleate is the maleate salt of thioperamide, a selective histamine H3 receptor antagonist used as a pharmacological tool to study histaminergic signaling. It is brain-penetrant and orally active, with a reported Ki of approximately 4.3 nM for H3 receptor inhibition.
  • Selective H3 receptor antagonist with Ki ≈ 4.3 nM.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • Provided as a solid, commonly supplied in 10 mg vials for research use.
  • High purity (>98%) as determined by vendor quality control.