Learn More
Cayman Chemical Thioperamide (maleate), Purity: ≥98%, CAS: 148440-81-7, 1 mg
SDP
Catalog No. 502887288
Encompass_Preferred
A histamine H3 receptor antagonist (Ki = 4.3 nM); selective for the histamine H3 receptor over histamine H1 and H2 receptors (Kis = >10,000 nM for both) and a panel of 36 other receptors and ion channels; has anticonvulsant activity, reverses immepip-induced antinociception, and promotes wakefulness in rats; decreases cognitive impairments in APP/PS1 mice
Catalog No. 50-288-7288 Supplier Cayman Chemical Supplier No. 100111271 MG
A histamine H3 receptor antagonist (Ki = 4.3 nM); selective for the histamine H3 receptor over histamine H1 and H2 receptors (Kis = >10,000 nM for both) and a panel of 36 other receptors and ion channels; has anticonvulsant activity, reverses immepip-induced antinociception, and promotes wakefulness in rats; decreases cognitive impairments in APP/PS1 mice