Learn More
Medchemexpress LLC Thioperoxydicarbonic acid ([(HO)C(S)]2S2), OC, OC'-diethyl ester | 502-55-6 | 98.0% | 100 MG
SDP
Catalog No. 5000412096
Encompass_Preferred
Dixanthogen is a dimer formed by the oxidation of xanthate. It can indirectly reflect the residual level of xanthates in water bodies. Xanthates are commonly used collectors in flotation processes but are highly toxic to the aquatic environment.
  • Dimer formed by oxidation of xanthate
  • Can indirectly reflect residual xanthate levels in water
  • For research use only
Catalog No. 50-004-12096 Supplier Medchemexpress LLC Supplier No. HYB1186100MG
Dixanthogen is a dimer formed by the oxidation of xanthate. It can indirectly reflect the residual level of xanthates in water bodies. Xanthates are commonly used collectors in flotation processes but are highly toxic to the aquatic environment.
  • Dimer formed by oxidation of xanthate
  • Can indirectly reflect residual xanthate levels in water
  • For research use only