Learn More
Medchemexpress LLC Thiophanat-ethyl-d10 | 1398066-16-4 | 380.51 | C14H8D10N4O4S2 | 10 MG
SDP
Catalog No. 5000281120
Encompass_Preferred
Thiophanat-ethyl-d10 is the deuterium-labeled isotopologue of thiophanat-ethyl provided for research and analytical applications. Supplied as a solid in a 10 mg quantity, it is intended for use as a stable-isotope internal standard and tracer to support quantitative methods. Consult the product datasheet and safety documentation for handling and storage information.
  • Deuterium-labeled (d10) isotopologue suitable as an internal standard.
  • Used as a tracer for quantitative NMR, GC-MS, and LC-MS analyses.
  • Provided as a solid, convenient for preparation of standards and calibrants.
  • Supports accurate quantitation by compensating for matrix effects and instrument variability.
  • Intended for research use; follow safety and storage recommendations on the datasheet.
Catalog No. 50-002-81120 Supplier Medchemexpress LLC Supplier No. HYW414878S10MG
Thiophanat-ethyl-d10 is the deuterium-labeled isotopologue of thiophanat-ethyl provided for research and analytical applications. Supplied as a solid in a 10 mg quantity, it is intended for use as a stable-isotope internal standard and tracer to support quantitative methods. Consult the product datasheet and safety documentation for handling and storage information.
  • Deuterium-labeled (d10) isotopologue suitable as an internal standard.
  • Used as a tracer for quantitative NMR, GC-MS, and LC-MS analyses.
  • Provided as a solid, convenient for preparation of standards and calibrants.
  • Supports accurate quantitation by compensating for matrix effects and instrument variability.
  • Intended for research use; follow safety and storage recommendations on the datasheet.