Learn More
Medchemexpress LLC Thiophanat-ethyl-d10 | 1398066-16-4 | 98.0% | 380.5 g·mol^-1 | C14H18N4O4S2 | 1 MG
SDP
Catalog No. 5000281038
Encompass_Preferred
Thiophanat-ethyl-d10 is the deuterium-labeled analogue of thiophanat-ethyl, provided as an isotopically labeled reference standard for analytical and research applications. Supplied in microgram/milligram quantities, it is intended for laboratory method development, calibration, and quantitative mass spectrometry.
  • Deuterium-labeled internal standard for mass spectrometry and chromatography.
  • High isotopic enrichment (≈99 atom % D) and minimum 98.0% chemical purity.
  • Supplied as a small-mass vial suitable for trace-level analytical work.
  • Applicable for method development, calibration, and quantitative analysis.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-81038 Supplier Medchemexpress LLC Supplier No. HYW414878S1MG
Thiophanat-ethyl-d10 is the deuterium-labeled analogue of thiophanat-ethyl, provided as an isotopically labeled reference standard for analytical and research applications. Supplied in microgram/milligram quantities, it is intended for laboratory method development, calibration, and quantitative mass spectrometry.
  • Deuterium-labeled internal standard for mass spectrometry and chromatography.
  • High isotopic enrichment (≈99 atom % D) and minimum 98.0% chemical purity.
  • Supplied as a small-mass vial suitable for trace-level analytical work.
  • Applicable for method development, calibration, and quantitative analysis.
  • For research use only; not for human or clinical applications.