Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiophanat-ethyl-d10 | 1398066-16-4 | 98.0% | 380.51 | C14H8D10N4O4S2 | 5 MG
SDP

Catalog No. 5000281371
Encompass_Preferred

Thiophanat-ethyl-d10 is the deuterium-labeled (d10) isotopologue of thiophanat-ethyl provided as a powder reference standard for analytical and research use. It is intended as an internal standard or tracer in quantitative, metabolic, and pharmacokinetic studies and should be stored per recommended conditions.

  • Deuterium-labeled d10 isotopologue.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Intended for use as an isotope-labeled reference standard and tracer.
  • Molecular formula C14H8D10N4O4S2; molecular weight 380.51.
  • Suitable for quantitative and metabolic/pharmacokinetic workflows.

Catalog No. 50-002-81371 Supplier Medchemexpress LLC Supplier No. HYW414878S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiophanat-ethyl-d10 is the deuterium-labeled (d10) isotopologue of thiophanat-ethyl provided as a powder reference standard for analytical and research use. It is intended as an internal standard or tracer in quantitative, metabolic, and pharmacokinetic studies and should be stored per recommended conditions.

  • Deuterium-labeled d10 isotopologue.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Intended for use as an isotope-labeled reference standard and tracer.
  • Molecular formula C14H8D10N4O4S2; molecular weight 380.51.
  • Suitable for quantitative and metabolic/pharmacokinetic workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.