Learn More
Medchemexpress LLC Thiophene-2 | 5MG
SDP
Catalog No. 5000229381
Encompass_Preferred
Thiophene-2 (TP2) is a specific polyketide synthase 13 (Pks13) inhibitor Thiophene-2 inhibits mycolic acid biosynthesis and rapidly leads to mycobacterial cell death Thiophene-2 is active against Mycobacterium tuberculosis with a MIC value of 1 M and has potent anti-tuberculosis activity[1]
Catalog No. 50-002-29381 Supplier Medchemexpress LLC Supplier No. HY1171455MG
Thiophene-2 (TP2) is a specific polyketide synthase 13 (Pks13) inhibitor Thiophene-2 inhibits mycolic acid biosynthesis and rapidly leads to mycobacterial cell death Thiophene-2 is active against Mycobacterium tuberculosis with a MIC value of 1 M and has potent anti-tuberculosis activity[1]