Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC13004EB
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC13004EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC13004EB Supplier Thermo Scientific Chemicals Supplier No. CC13004EB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-03-3
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.15
MDL Number MFCD00005429
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
PubChem CID 7364
ChEBI CHEBI:87301
IUPAC Name thiophene-2-carbaldehyde
SMILES O=CC1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 112.15
Percent Purity 97%
Chemical Name or Material Thiophene-2-carbaldehyde
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.