Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thiophene-2-carbonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Density | 1.171 |
Boiling Point | 191°C to 193°C |
Flash Point | 81°C (178°F) |
Odor | Unpleasant |
Refractive Index | 1.5645 |
Quantity | 500 g |
UN Number | UN3276 |
Beilstein | 107035 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 109.15 |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H315-H318-H335 |
Hazard Statement | GHS H Statement H301-H227-H312-H332-H315-H319 Toxic if swallowed. Combustible liquid. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: III |
EINECSNumber | 213-706-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction