Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-2-carbothioamide 98.0+%, TCI America™
SDP

Catalog No. T36045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36045G 5 g
T360425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36045G Supplier TCI America Supplier No. T36045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20300-02-1
Molecular Formula C5H5NS2
Molecular Weight (g/mol) 143.222
MDL Number MFCD00052890
InChI Key HDXYHAPUCGQOBX-UHFFFAOYSA-N
Synonym thiophene-2-thiocarboxamide, 2-thiophenecarbothioamide, thiophene-2-thioamide, thiophene-2-carbothioic acid amide, thiophene,2-thiocarboxamide, amino-2-thienylmethane-1-thione, 2-thienylthioamide, 2-carbamothioylthiophene, acmc-1cfey, 2-thiophenethiocarboxamide
PubChem CID 737237
IUPAC Name thiophene-2-carbothioamide
SMILES C1=CSC(=C1)C(=S)N

Specifications

Melting Point 103°C
Color Yellow
Quantity 5 g
Formula Weight 143.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Thiophene-2-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.