Learn More
Matrix Scientific Thiophene-3-boronic acid, 6165-69-1, MFCD00151851, 5g
Supplier: Matrix Scientific 0014535G
Molecular Formula C4H5BO2S, Purity 98%, Molecular Weight 127.96, Melting Point 166-168°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.