Learn More
Thiophene-3-carboxylic acid, 99%, Thermo Scientific Chemicals
Description
3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.
Solubility
Soluble in water at 20°C 4.3 g/L.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 137°C to 141°C |
Quantity | 5 g |
Beilstein | 1994 |
Merck Index | 14,9279 |
Solubility Information | Soluble in water at 20°C 4.3g/L. |
Formula Weight | 128.15 |
Percent Purity | 99% |
Chemical Name or Material | Thiophene-3-carboxylic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 201-802-5 |
RTECSNumber | XM8330300 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.