Learn More
Medchemexpress LLC Thiophene-3-carboxylic acid derivative | 509083-77-6 | 99.4% | 50 MG
SDP
Catalog No. 5000232248
Encompass_Preferred
BMS-587101 is a potent, orally active small-molecule antagonist of leukocyte function-associated antigen-1 (LFA-1) used in preclinical immunology and inflammation research. Supplied as a purified solid for laboratory use.
  • Potent LFA-1 antagonist for immunology research.
  • Demonstrated anti-inflammatory activity in preclinical models.
  • Chemical formula C26H20Cl2N4O4S; molecular weight 555.43 g·mol⁻¹.
  • High purity (99.4%) suitable for research applications.
  • Available in small solid quantities (for example, 50 MG) for laboratory experiments.
Catalog No. 50-002-32248 Supplier Medchemexpress LLC Supplier No. HY12062850MG
BMS-587101 is a potent, orally active small-molecule antagonist of leukocyte function-associated antigen-1 (LFA-1) used in preclinical immunology and inflammation research. Supplied as a purified solid for laboratory use.
  • Potent LFA-1 antagonist for immunology research.
  • Demonstrated anti-inflammatory activity in preclinical models.
  • Chemical formula C26H20Cl2N4O4S; molecular weight 555.43 g·mol⁻¹.
  • High purity (99.4%) suitable for research applications.
  • Available in small solid quantities (for example, 50 MG) for laboratory experiments.