Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiophene-3-carboxylic acid triazaspiro nonan derivative | 509083-77-6 | 99.4% | 25 MG
SDP

Catalog No. 5000234869
Encompass_Preferred

Small-molecule LFA-1 antagonist (CAS 509083-77-6) supplied as a high-purity research reagent for in vitro and in vivo pharmacology studies.

  • High purity: 99.4% (LCMS, rounded).
  • Molecular weight: 555.43 g/mol.
  • Chemical formula: C26H20Cl2N4O4S.
  • Suitable for pharmacology and preclinical inflammation models.
  • Available in small laboratory pack sizes for research use.

Catalog No. 50-002-34869 Supplier Medchemexpress LLC Supplier No. HY12062825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule LFA-1 antagonist (CAS 509083-77-6) supplied as a high-purity research reagent for in vitro and in vivo pharmacology studies.

  • High purity: 99.4% (LCMS, rounded).
  • Molecular weight: 555.43 g/mol.
  • Chemical formula: C26H20Cl2N4O4S.
  • Suitable for pharmacology and preclinical inflammation models.
  • Available in small laboratory pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.