Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thiophene

Catalog No. T31801500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31801500G 500 g
T318015G 5 g
T31801100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T31801500G Supplier Sigma Aldrich Supplier No. T31801500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thiophene

Specifications

CAS 110-02-1
Melting Point -38°C (lit.)
Density 1.051 g/mL (at 25°C (literature))
Boiling Point 84°C (lit.)
Molecular Formula C4H4S
Linear Formula C4H4S
Refractive Index n20/D 1.529 (literature)
MDL Number MFCD00005413
Quantity 500 g
Synonym Thiofuran; Thiole
Molecular Weight (g/mol) 84.14
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.