Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophenol, 99%, AcroSeal™

Catalog No. p-4229342
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430721000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC430721000 Supplier Thermo Scientific Chemicals Supplier No. 430721000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 108-98-5 | C6H6S | 110.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-98-5
Molecular Formula C6H6S
Molecular Weight (g/mol) 110.17
MDL Number MFCD00004826
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
PubChem CID 7969
ChEBI CHEBI:48498
IUPAC Name benzenethiol
SMILES C1=CC=C(C=C1)S

Specifications

Melting Point -15.0°C
Color Colorless to Yellow
Density 1.0780g/mL
Boiling Point 167.0°C to 169.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5880 to 1.5900
Linear Formula C6H5SH
Quantity 100 mL
Merck Index 15,9508
Specific Gravity 1.078
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2
Viscosity 0.981 mm2/s (30°C)
Formula Weight 110.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Suspected of damaging fertility or the unbo
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou
EINECSNumber 203-635-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.