Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophenol, 99+%

Catalog No. AAA1591609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1591609 10 g
AAA1591618 50 g
AAA1591630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1591609 Supplier Thermo Scientific Chemicals Supplier No. A1591609

CAS: 108-98-5 | C6H6S | 110.174 g/mol

Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.

Notes
Incompatible with strong acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-98-5
Molecular Formula C6H6S
Molecular Weight (g/mol) 110.174
MDL Number MFCD00004826
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto
PubChem CID 7969
ChEBI CHEBI:48498
IUPAC Name benzenethiol
SMILES C1=CC=C(C=C1)S

Specifications

Melting Point -15°C
Density 1.075
pH 5
Boiling Point 168°C to 169°C
Flash Point 55°C (131°F)
Odor Stench
Assay Percent Range ≥99%
Refractive Index 1.59
Quantity 10 g
UN Number UN2337
Beilstein 506523
Sensitivity Air Sensitive
Merck Index 14,9355
Solubility Information Miscible with most organic solvents and aqueous base. Immiscible with water.
Formula Weight 110.18
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less
Hazard Category H226-H300+H310+H330-H315-H319-H335-H361-H371-H372
Hazard Statement GHS H Statement
H300-H310-H330-H318-H226-H361-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280i-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P37
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHENYL MERCAPTAN
EINECSNumber 203-635-3
RTECSNumber DC0525000
TSCA Yes
storageNote1 Air sensitive
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.