Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophenol, Approx. 97%, Spectrum™ Chemical
SDP

Catalog No. 18607385 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
50 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-385 50 kg Amber Glass Bottle
18-607-384 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-385 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T226050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T2260, 108-98-5, C6H6S

Spectrum™ Chemical Thiophenol , also known as Benzenethiol or Phenyl mercaptan, is an organosulfur compound and colorless liquid which is considered the simplest aromatic thiol. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

CAS 108-98-5
CAS Max % 1
Boiling Point 168 to 169°C
Packaging Amber Glass Bottle
Quantity 50 kg
Percent Purity ∼97%
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.