Learn More
Bioworld THIOPHILIC SEPAROPOREAGAR
Supplier:  Bioworld 201810383
Encompass
THIOPHILIC SEPAROPORE (AGAROSE) 4B-CL: The term Thiophilic refers to an affinity for sulfone groups that lie in close proximity to thioether groups. Thiophilic affinity purification is based on the ability of some proteins, particularl...
Catalog No. 50-197-762