Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thiophosphoro Alkyne Dichloridate

Catalog No. 914746100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-474-6100MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-474-6100MG Supplier Sigma Aldrich Supplier No. 914746100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thiophosphoro alkyne dichloridate (TPAC) is a histidine labeling reagent developed by the Chang lab that mimics native post-translational His phosphorylation. The alkyne allows for subsequent reactions or addition of payloads by copper-assisted alkyne-azide cycloaddition (CuAAC) chemistry. TPAC was also demonstrated to covalently modify His-tags, and by clicking on polyarginine tags, enabled protein delivery into live cells.Bioinspired Thiophosphorodichloridate Reagents for Chemoselective Histidine Bioconjugation

Specifications

CAS 150571-28-1
Molecular Formula C40H65FN6O6
Quantity 50 mg
Synonym tert-Butyl ((S)-1-(((S)-2-((S)-2-(((S)-1-((10-Aminodecyl)amino)-3-(4-fluorophenyl)-1-oxopropan-2-yl)carbamoyl)pyrrolidin-1-yl)-1-cyclohexyl-2-oxoethyl)amino)-1-oxopropan-2-yl)carbamate; AVP conjugate for IAP-mediated protein degrader development; SNIPER building block
Molecular Weight (g/mol) 744.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.