Learn More
Strem, An Ascensus Company CAS# 3982-91-0. 100g. Thiophosphoryl chloride, 98%. MFCD00011504
SDP
Catalog No. 5090114245
Encompass
CAS# 3982-91-0. 100g. Thiophosphoryl chloride, 98%. MFCD00011504. Molecular Weight: 169.40. Molecular Formula: PSCl3. Color/form: colorless liq. Strem# 93-1591. http://www.strem.com/catalog/v/93-1591/
Catalog No. 50-901-14245 Supplier Strem, An Ascensus Company Supplier No. 931591100G
CAS# 3982-91-0. 100g. Thiophosphoryl chloride, 98%. MFCD00011504. Molecular Weight: 169.40. Molecular Formula: PSCl3. Color/form: colorless liq. Strem# 93-1591. http://www.strem.com/catalog/v/93-1591/