Learn More
Cayman Chemical Thiopropamine (hydrochloride), Purity: ≥98%, CAS: 53632-92-1, 10 mg
SDP
Catalog No. 502895569
Encompass_Preferred
An analog of amphetamine in which the phenyl ring has been replaced by a thiophene group
Catalog No. 50-289-5569 Supplier Cayman Chemical Supplier No. 900203710 MG
An analog of amphetamine in which the phenyl ring has been replaced by a thiophene group