Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thioquinapiperifil dihydrochloride | 204077-66-7 | 99.2% | 1 ML
SDP

Catalog No. 5000233097
Encompass_Preferred

Thioquinapiperifil dihydrochloride is a potent, selective, non-competitive phosphodiesterase-5 (PDE5) inhibitor supplied as a 10 mM solution in DMSO for research use. It is provided for studies of PDE5 modulation and related sexual function research.

  • Ready-to-use 10 mM solution in DMSO
  • High purity at 99.22%
  • Molecular formula C24H30Cl2N6OS; molecular weight 521.51 g·mol⁻¹
  • Potent PDE5 inhibition (IC50 0.074 nM)

Catalog No. 50-002-33097 Supplier Medchemexpress LLC Supplier No. HY119611A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thioquinapiperifil dihydrochloride is a potent, selective, non-competitive phosphodiesterase-5 (PDE5) inhibitor supplied as a 10 mM solution in DMSO for research use. It is provided for studies of PDE5 modulation and related sexual function research.

  • Ready-to-use 10 mM solution in DMSO
  • High purity at 99.22%
  • Molecular formula C24H30Cl2N6OS; molecular weight 521.51 g·mol⁻¹
  • Potent PDE5 inhibition (IC50 0.074 nM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.