Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thioquinapiperifil dihydrochloride | 204077-66-7 | 99.2% | 100 MG
SDP

Catalog No. 5000233598
Encompass_Preferred

Thioquinapiperifil dihydrochloride (KF31327) is a potent, selective, non-competitive phosphodiesterase-5 (PDE-5) inhibitor (reported IC50 0.074 nM) supplied for laboratory research. It is provided as a purified solid and as a DMSO solution, with solubility and storage guidance for in vitro and in vivo studies.

  • Potent selective PDE-5 inhibitor (reported IC50 0.074 nM).
  • Available as solid and 10 mM DMSO solution.
  • High purity (~99.2%) suitable for research applications.
  • Molecular weight 521.51 g/mol; formula C24H30Cl2N6OS.
  • Soluble in DMSO; in vivo formulation protocols provided.
  • Store solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-33598 Supplier Medchemexpress LLC Supplier No. HY119611A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thioquinapiperifil dihydrochloride (KF31327) is a potent, selective, non-competitive phosphodiesterase-5 (PDE-5) inhibitor (reported IC50 0.074 nM) supplied for laboratory research. It is provided as a purified solid and as a DMSO solution, with solubility and storage guidance for in vitro and in vivo studies.

  • Potent selective PDE-5 inhibitor (reported IC50 0.074 nM).
  • Available as solid and 10 mM DMSO solution.
  • High purity (~99.2%) suitable for research applications.
  • Molecular weight 521.51 g/mol; formula C24H30Cl2N6OS.
  • Soluble in DMSO; in vivo formulation protocols provided.
  • Store solid at 4°C; solutions at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.