Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thioquinapiperifil dihydrochloride | 204077-66-7 | 99.2% | 5 MG
SDP

Catalog No. 5000233654
Encompass_Preferred

Thioquinapiperifil dihydrochloride is a potent, selective, non-competitive phosphodiesterase-5 (PDE-5) inhibitor (reported IC50 0.074 nM) supplied as the dihydrochloride salt for research use in PDE-5 pharmacology.

  • Potent, selective, non-competitive PDE-5 inhibitor (reported IC50 0.074 nM).
  • Dihydrochloride salt form for improved handling and solubility.
  • High purity suitable for research use.
  • Well-defined chemical identity: formula C24H30Cl2N6OS, MW 521.51 g·mol⁻1.
  • Available in small research pack sizes, including 5 mg solid and a 10 mM solution format.

Catalog No. 50-002-33654 Supplier Medchemexpress LLC Supplier No. HY119611A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thioquinapiperifil dihydrochloride is a potent, selective, non-competitive phosphodiesterase-5 (PDE-5) inhibitor (reported IC50 0.074 nM) supplied as the dihydrochloride salt for research use in PDE-5 pharmacology.

  • Potent, selective, non-competitive PDE-5 inhibitor (reported IC50 0.074 nM).
  • Dihydrochloride salt form for improved handling and solubility.
  • High purity suitable for research use.
  • Well-defined chemical identity: formula C24H30Cl2N6OS, MW 521.51 g·mol⁻1.
  • Available in small research pack sizes, including 5 mg solid and a 10 mM solution format.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.