Learn More
Cayman Chemical ThIordoxIn 1 RabIt MoncLN 50uL
SDP
Catalog No. 502915231
Encompass_Preferred
A peptide TRPV6 antagonist inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 14 nM) reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model
Catalog No. 50-291-5231 Supplier Cayman Chemical Supplier No. 3707350 UL
A peptide TRPV6 antagonist inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 14 nM) reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model