Learn More
Enzo Life Sciences Thioredoxin monoclonal antibody (2C9) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIMSA150E
Encompass_Preferred
Clone: 2C9. Host: Mouse. Isotype: IgG1. Immunogen: E. coli thioredoxin (trxA). Species reactivity: E. coli. Specificity: Recognizes trx and trx-tag-f thioredoxin. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7349