Learn More
Medchemexpress LLC HY-P1948A 1mg Medchemexpress, Thioredoxin reductase peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1948A1MG
Encompass_Preferred
Medchemexpress, HY-P1948A 1mg Thioredoxin reductase peptide (TFA) CAS: Thioredoxin reductase peptide TFA corresponds to residues 53–67 in thioredoxin reductase (TrxR), used in thioredoxin reductase research. Thioredoxin reductase acts as a reductant of disulfide-containing proteins and plays crucial role in cellular antioxidant defense [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4253