Learn More
Medchemexpress LLC Thioridazine-d3 hydrochloride | 1189928-36-6 | 98.3% | 1 MG
SDP
Catalog No. 5000408027
Encompass_Preferred
Thioridazine-d3 hydrochloride is the deuterium-labeled version of Thioridazine, which is an antagonist of dopamine receptor D2 family proteins. It demonstrates potent anti-psychotic and anti-anxiety effects. Additionally, Thioridazine is a strong inhibitor of PI3K-Akt-mTOR signaling pathways, exhibiting anti-angiogenic properties and showing antiproliferative and apoptosis induction in various cancer cell types, particularly targeting cancer stem cells (CSCs).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08027 Supplier Medchemexpress LLC Supplier No. HYB0965AS1MG
Thioridazine-d3 hydrochloride is the deuterium-labeled version of Thioridazine, which is an antagonist of dopamine receptor D2 family proteins. It demonstrates potent anti-psychotic and anti-anxiety effects. Additionally, Thioridazine is a strong inhibitor of PI3K-Akt-mTOR signaling pathways, exhibiting anti-angiogenic properties and showing antiproliferative and apoptosis induction in various cancer cell types, particularly targeting cancer stem cells (CSCs).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.