Learn More
Apexbio Technology LLC Thioridazine HCl,100mg CAS# 130-61-0
SDP
Catalog No. 502240620
Encompass_Preferred
Thioridazine·HCl is a calcium channel protein inhibitor as well as D2DR/D4DR inhibitor. Its administration has been associated with hypotension. It has also been, after chronic treatment, used to test increased sensitivity of dopaminergic agents. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0620 Supplier Apexbio Technology LLC Supplier No. A8537100
Thioridazine·HCl is a calcium channel protein inhibitor as well as D2DR/D4DR inhibitor. Its administration has been associated with hypotension. It has also been, after chronic treatment, used to test increased sensitivity of dopaminergic agents. Other sizes are also available. Please inqury us for quote.