Learn More
Apexbio Technology LLC Thioridazine HCl 130-61-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061494
Encompass_Preferred
Thioridazine HCl is a pharmacological inhibitor targeting dopamine receptor subtypes D2 and D4 as well as a calcium channel blocker It modulates dopaminergic neurotransmission by interfering with receptor-mediated signaling and reduces intracellular calcium influx via calcium channel inhibition (IC50 approximately 1 0 M) Thioridazine is frequently utilized in research contexts involving dopaminergic signaling pathways calcium-related cellular processes and investigations of pharmacological sensitivity shifts following prolonged receptor modulation Specifically it is employed experimentally to examine receptor-mediated cardiovascular responses such as hypotension and to assess alterations in dopamine receptor responsiveness induced by chronic exposure regimens
Catalog No. 50-000-61494 Supplier Apexbio Technology LLC Supplier No. A85375.1
Thioridazine HCl is a pharmacological inhibitor targeting dopamine receptor subtypes D2 and D4 as well as a calcium channel blocker It modulates dopaminergic neurotransmission by interfering with receptor-mediated signaling and reduces intracellular calcium influx via calcium channel inhibition (IC50 approximately 1 0 M) Thioridazine is frequently utilized in research contexts involving dopaminergic signaling pathways calcium-related cellular processes and investigations of pharmacological sensitivity shifts following prolonged receptor modulation Specifically it is employed experimentally to examine receptor-mediated cardiovascular responses such as hypotension and to assess alterations in dopamine receptor responsiveness induced by chronic exposure regimens