Learn More
Medchemexpress LLC Thioridazine hydrochloride | 130-61-0 | 99.96% | 100 MG
SDP
Catalog No. 5000409097
Encompass_Preferred
Thioridazine hydrochloride is an orally active antagonist of dopamine receptor D2 family proteins, demonstrating potent anti-psychotic and anti-anxiety activities. It also acts as a powerful inhibitor of PI3K-Akt-mTOR signaling pathways, exhibiting anti-angiogenic effects. This compound shows antiproliferative and apoptosis induction effects in various cancer cell types, with a specific focus on targeting cancer stem cells (CSCs).
  • Orally active.
  • Dopamine receptor D2 family protein antagonist.
  • Potent anti-psychotic activity.
  • Potent anti-anxiety activity.
  • Potent inhibitor of PI3K-Akt-mTOR signaling pathways.
  • Anti-angiogenic effect.
  • Antiproliferative effects in various cancer cell types.
  • Apoptosis induction effects in various cancer cell types.
  • Specificity in targeting cancer stem cells (CSCs).
Catalog No. 50-004-09097 Supplier Medchemexpress LLC Supplier No. HYB0965100MG
Add to Cart
Add to Cart
Thioridazine hydrochloride is an orally active antagonist of dopamine receptor D2 family proteins, demonstrating potent anti-psychotic and anti-anxiety activities. It also acts as a powerful inhibitor of PI3K-Akt-mTOR signaling pathways, exhibiting anti-angiogenic effects. This compound shows antiproliferative and apoptosis induction effects in various cancer cell types, with a specific focus on targeting cancer stem cells (CSCs).
  • Orally active.
  • Dopamine receptor D2 family protein antagonist.
  • Potent anti-psychotic activity.
  • Potent anti-anxiety activity.
  • Potent inhibitor of PI3K-Akt-mTOR signaling pathways.
  • Anti-angiogenic effect.
  • Antiproliferative effects in various cancer cell types.
  • Apoptosis induction effects in various cancer cell types.
  • Specificity in targeting cancer stem cells (CSCs).