Learn More
Medchemexpress LLC Thioridazine hydrochloride | 130-61-0 | 99.96% | 500 MG
SDP
Catalog No. 5000408107
Encompass_Preferred
Thioridazine hydrochloride is an orally active antagonist of dopamine receptor D2 family proteins, showing potent anti-psychotic and anti-anxiety activities. It is also a potent inhibitor of PI3K-Akt-mTOR signaling pathways with anti-angiogenic effects. Additionally, it demonstrates antiproliferative and apoptosis induction in various cancer cells, specifically targeting cancer stem cells (CSCs).
  • Potent anti-psychotic and anti-anxiety activities
  • Inhibitor of PI3K-Akt-mTOR signaling pathways
  • Anti-angiogenic effects
  • Antiproliferative and apoptosis induction in cancer cells
  • Specifically targets cancer stem cells (CSCs)
Catalog No. 50-004-08107 Supplier Medchemexpress LLC Supplier No. HYB0965500MG
Thioridazine hydrochloride is an orally active antagonist of dopamine receptor D2 family proteins, showing potent anti-psychotic and anti-anxiety activities. It is also a potent inhibitor of PI3K-Akt-mTOR signaling pathways with anti-angiogenic effects. Additionally, it demonstrates antiproliferative and apoptosis induction in various cancer cells, specifically targeting cancer stem cells (CSCs).
  • Potent anti-psychotic and anti-anxiety activities
  • Inhibitor of PI3K-Akt-mTOR signaling pathways
  • Anti-angiogenic effects
  • Antiproliferative and apoptosis induction in cancer cells
  • Specifically targets cancer stem cells (CSCs)