Learn More
Medchemexpress LLC Thiosalicylic acid | 147-93-3 | MFCD00004836 | 99.3% | 154.19 | C7H6O2S | 500g
SDP
Catalog No. 5000268883
Encompass_Preferred
Thiosalicylic acid (2-Mercaptobenzoic acid) is a stabilizer that can be used in the synthesis of metal nanoparticles Thiosalicylic acid is a biomaterial or organic compound that can be used in life science research[1]
Catalog No. 50-002-68883 Supplier Medchemexpress LLC Supplier No. HYY0102500G
Thiosalicylic acid (2-Mercaptobenzoic acid) is a stabilizer that can be used in the synthesis of metal nanoparticles Thiosalicylic acid is a biomaterial or organic compound that can be used in life science research[1]