Learn More
Electron Microscopy Sciences Thiosemicarbazide 25 GR

Supplier: Electron Microscopy Sciences 22000
CAS #79-19-6 NH₂CSNHNH₂ F.W. 91.13
Assay: >99.0%
Melting point: 181-184 °C (358-363 °F)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.