Learn More
Medchemexpress LLC Thiosemicarbazide | 79-19-6 | MFCD00007620 | >98.0% | 91.14 g/mol | CH5N3S | 10g
SDP
Catalog No. 5000270182
Encompass_Preferred
Thiosemicarbazide is a vitamin B6 antagonist with anti-acne activity Thiosemicarbazide is also a well-known source in the synthesis of heterocycles and its derivatives have potential anticancer activity Thiosemicarbazide (TSC HL1) reacts with metal salts urea (U) to prepare Co(II) and Cu(I) metal complexes Thiosemicarbazide is also used in the fields of media communications and optical storage and in the spectrophotometric detection of metals[1][2][3][1][2][3]
Catalog No. 50-002-70182 Supplier Medchemexpress LLC Supplier No. HYY003210G
Thiosemicarbazide is a vitamin B6 antagonist with anti-acne activity Thiosemicarbazide is also a well-known source in the synthesis of heterocycles and its derivatives have potential anticancer activity Thiosemicarbazide (TSC HL1) reacts with metal salts urea (U) to prepare Co(II) and Cu(I) metal complexes Thiosemicarbazide is also used in the fields of media communications and optical storage and in the spectrophotometric detection of metals[1][2][3][1][2][3]