Learn More
Medchemexpress LLC Thiosemicarbazide | 79-19-6 | MFCD00007620 | >=98.0% | 91.14 g/mol | CH5N3S | 1mL
SDP
Catalog No. 5000269089
Encompass_Preferred
Thiosemicarbazide is a vitamin B6 antagonist with anti-acne activity Thiosemicarbazide is also a well-known source in the synthesis of heterocycles and its derivatives have potential anticancer activity Thiosemicarbazide (TSC HL1) reacts with metal salts urea (U) to prepare Co(II) and Cu(I) metal complexes Thiosemicarbazide is also used in the fields of media communications and optical storage and in the spectrophotometric detection of metals[1][2][3][1][2][3]
Catalog No. 50-002-69089 Supplier Medchemexpress LLC Supplier No. HYY003210MM/1ML
Thiosemicarbazide is a vitamin B6 antagonist with anti-acne activity Thiosemicarbazide is also a well-known source in the synthesis of heterocycles and its derivatives have potential anticancer activity Thiosemicarbazide (TSC HL1) reacts with metal salts urea (U) to prepare Co(II) and Cu(I) metal complexes Thiosemicarbazide is also used in the fields of media communications and optical storage and in the spectrophotometric detection of metals[1][2][3][1][2][3]