Learn More
Enzo Life Sciences Thiostrepton (5g). CAS: 1393-48-2
SDP
Supplier:  Enzo Life Sciences ALX380261G005
Encompass_Preferred
Thiazole-containing peptide antibiotic. Inhibits protein synthesis by preventing binding of GTP to the 50S ribosomal subunit. Inhibits binding of elongation factor G (EF-G) and the dissociation of EF-G from the ribosome. Solubility: Soluble in acetic acid, DMSO (10mg/ml) or dimethyl formamide; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8992