Learn More
Thiostrepton, G-Biosciences

Description
Thiostrepton has been shown to inhibit FOXM1, a gene commonly overexpressed in cancer cell lines, thus inhibiting cancer cell growth. Thiostrepton functions by binding elongation factor G (EF-G) and interfering with ribosomal GTPase activity, ultimately inhibiting protein synthesis.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 1 g |
| Product Line | Gibco |
| Percent Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.