Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Thiosulfate Citrate Bile Sucrose Agar, for Vibrio
SDP

Catalog No. 23001813 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-813 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-813 Supplier Hardy Diagnostics™ Supplier No. G55
Add to Cart
Add to Cart
This item is not returnable. View return policy

TCBS Agar (Thiosulfate Citrate Bile Salts Sucrose Agar) is recommended for the selective isolation and cultivation of Vibrio spp. from clinical specimens

  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Packaged in sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture

Specifications

Appearance Slightly opalescent with no precipitate
Color Dark Green
Content And Storage 2°C to 8°C
Detects Vibrio spp.
Form Agar
Format Agar Plate
Media Type TCBS Agar
Product Type Agar Plate
Target Organism Class Gram-negative Bacteria
Test Type Microbial Cultivation
Fill Volume 18 mL
Formulation Sucrose - 20.0gm, Dipeptone - 10.0gm, Sodium Citrate - 10.0gm, Sodium Thiosulfate - 10.0gm, Sodium Chloride - 10.0gm, Yeast Extract - 5.0gm, Oxbile (Oxgall) - 5.0gm, Sodium Cholate - 3.0gm, Ferric Citrate - 1.0gm, Bromothymol Blue - 0.04gm, Thymol Blue - 0.04gm, Agar - 15.0gm
For Use With (Application) Selective isolation and cultivation of Vibrio spp.
Incubation Conditions 35°C
Incubation Time 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Vibrio parahaemolyticus
Protein Source Dipeptone, Yeast Extract, Oxgall
Source Plant, Animal, Yeast
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.