Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thiourea

Catalog No. T787512KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
12 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T787512KG 12 kg
T78751KG 1 kg
T78755KG 5 kg
T78755G 5 g
T7875100G 100 g
T7875500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. T787512KG Supplier Sigma Aldrich Supplier No. T787512KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thiourea

Specifications

CAS 62-56-6
Melting Point 170°C to 176°C
Molecular Formula CH4N2S
Linear Formula NH2CSNH2
Quantity 12 kg
MDL Number MFCD00008067
Synonym Sulfourea; Thiocarbamide
Molecular Weight (g/mol) 76.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.