Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thiourea

Catalog No. T7875500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
12 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T7875500G 500 g
T787512KG 12 kg
T78751KG 1 kg
T78755KG 5 kg
T78755G 5 g
T7875100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. T7875500G Supplier Sigma Aldrich Supplier No. T7875500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thiourea

Specifications

CAS 5927-18-4
Density 1.125 g/mL at 25°C
Boiling Point 118°C (0.85 mmHg, lit.), 265°C to 268°C
Molecular Formula C5H11O5P
Linear Formula (CH3O)2P(O)CH2CO2CH3
Quantity 500 g
MDL Number MFCD00008452
Synonym Sulfourea; Thiocarbamide
Molecular Weight (g/mol) 182.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.